Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabggtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138708.2 | NP_619715.1 | 4 - 5 | 483 | 107 | |
GenBank mRNA | BC060536.1 | - | 507 | 107 | ||
BC078683.1 | - | 4 - 5 | 472 | 7135106 | 107 | |
FQ214773.1 | - | 487 | 107 | |||
FQ216744.1 | - | 487 | 107 | |||
FQ224712.1 | - | 486 | 107 | |||
FQ229109.1 | - | 487 | 107 | |||
L10416.1 | - | 4 - 5 | 483 | 107 | ||
S62097.1 | - | 4 - 5 | 483 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |