Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zranb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031616.1 | NP_113804.1 | 2 - 3 | 120 | 64 | |
GenBank mRNA | AF013965.1 | - | 2 - 3 | 360 | 64 | |
AF013967.1 | - | 2 - 3 | 120 | 64 | ||
BC087012.1 | - | 2 - 3 | 106 | 7189430 | 64 | |
FQ211774.1 | - | 2 - 3 | 146 | 64 | ||
FQ211917.1 | - | 2 - 3 | 146 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |