Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022633.1 | NP_072155.1 | 2 - 3 | 206 | 84 | |
GenBank mRNA | AF188491.1 | - | 2 - 3 | 206 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |