Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173148.1 | NP_775171.1 | 582 | 73 | ||
GenBank mRNA | M76734.1 | - | 582 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |