Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106134.1 | NP_001099604.1 | 4 - 5 | 506 | 96 | |
XM_006254918.2 | XP_006254980.1 | 4 - 5 | 519 | 96 | ||
XM_006254920.2 | XP_006254982.1 | 4 - 5 | 512 | 96 | ||
GenBank mRNA | BC158659.1 | - | 4 - 5 | 458 | 8369180 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |