Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cybb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023965.1 | NP_076455.1 | 11 - 12 | 1460 | 58 | |
GenBank mRNA | AF298656.3 | - | 11 - 12 | 1460 | 58 | |
AJ295950.1 | - | 11 - 12 | 1460 | 58 | ||
FQ217794.1 | - | 1529 | 58 | |||
FQ231233.1 | - | 1512 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |