Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031584.2 | NP_113772.1 | 3 - 4 | 762 | 69 | |
GenBank mRNA | D83044.1 | - | 3 - 4 | 765 | 69 | |
X98334.1 | - | 3 - 4 | 711 | 69 | ||
Y13154.1 | - | 3 - 4 | 712 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |