Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sft2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008302.1 | NP_001008303.1 | 3 - 4 | 297 | 88 | |
GenBank mRNA | BC085346.1 | - | 3 - 4 | 297 | 7304630 | 88 |
FQ225074.1 | - | 3 - 4 | 88 | |||
FQ227930.1 | - | 3 - 4 | 275 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |