Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008770.3 | NP_001008770.1 | 4 - 5 | 505 | 108 | |
XM_006232097.2 | XP_006232159.1 | 4 - 5 | 2616 | 108 | ||
GenBank mRNA | AY325197.1 | - | 4 - 5 | 507 | 108 | |
BC088459.1 | - | 3 - 4 | 489 | 7127481 | 108 | |
FQ209709.1 | - | 4 - 5 | 663 | 108 | ||
FQ213936.1 | - | 4 - 5 | 515 | 108 | ||
FQ215832.1 | - | 3 - 4 | 507 | 108 | ||
FQ219291.1 | - | 4 - 5 | 594 | 108 | ||
FQ219760.1 | - | 4 - 5 | 518 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |