Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760824.1 | XP_008759046.1 | 2251 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |