Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012928.1 | NP_037060.1 | 552 | 86 | ||
GenBank mRNA | AF126808.1 | - | 446 | 86 | ||
X55519.1 | - | 552 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Recommended Assay Rattus norvegicus Probe spans exons |