Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031630.2 | NP_113818.2 | 4 - 5 | 330 | 89 | |
GenBank mRNA | AF142629.2 | - | 4 - 5 | 330 | 89 | |
BC078791.1 | - | 4 - 5 | 312 | 7112991 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |