Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Barx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750457.3 | XP_003750505.1 | 1 - 2 | 740 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |