Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108131.1 | NP_001101601.1 | 7 - 8 | 2324 | 59 | |
XM_006242747.2 | XP_006242809.1 | 8 - 9 | 2641 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |