Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153302.1 | NP_695214.1 | 1 - 2 | 203 | 79 | |
GenBank mRNA | AF411249.1 | - | 1 - 2 | 203 | 79 | |
BC089773.1 | - | 1 - 2 | 198 | 79 | ||
BC105900.1 | - | 1 - 2 | 208 | 7377732 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |