Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034942.1 | NP_001030114.1 | 2 - 3 | 1173 | 79 | |
XM_006242768.2 | XP_006242830.1 | 2 - 3 | 1360 | 79 | ||
XM_006242769.2 | XP_006242831.1 | 2 - 3 | 1360 | 79 | ||
GenBank mRNA | BC104680.1 | - | 2 - 3 | 1173 | 7456912 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |