Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256694.1 | XP_006256756.1 | 6 - 7 | 609 | 148 | |
XM_006256695.1 | XP_006256757.1 | 6 - 7 | 606 | 148 | ||
GenBank mRNA | FQ210873.1 | - | 6 - 7 | 566 | 148 | |
FQ220834.1 | - | 6 - 7 | 562 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |