Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108247.2 | NP_001101717.1 | 2 - 3 | 361 | 115 | |
XM_006256719.2 | XP_006256781.1 | 3 - 4 | 902 | 115 | ||
XM_006256720.2 | XP_006256782.1 | 3 - 4 | 1159 | 115 | ||
XM_008773075.1 | XP_008771297.1 | 3 - 4 | 1130 | 115 | ||
GenBank mRNA | AF452730.1 | - | 3 - 4 | 1109 | 115 | |
BC167040.1 | - | 2 - 3 | 361 | 7135266 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |