Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108538.1 | NP_001102008.1 | 4 - 5 | 797 | 84 | |
XM_008773000.1 | XP_008771222.1 | 6 - 7 | 767 | 84 | ||
GenBank mRNA | BC166724.1 | - | 6 - 7 | 728 | 7933637 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |