Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165880.1 | NP_001159352.1 | 1 - 2 | 95 | 66 | |
XM_006255979.2 | XP_006256041.1 | 2 - 3 | 198 | 66 | ||
GenBank mRNA | BC127463.1 | - | 1 - 2 | 95 | 8362353 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |