Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017196.3 | NP_058892.1 | 2 - 3 | 184 | 89 | |
XM_006256061.2 | XP_006256123.1 | 2 - 3 | 279 | 89 | ||
XM_006256062.2 | XP_006256124.1 | 3 - 4 | 256 | 89 | ||
XM_006256063.2 | XP_006256125.1 | 3 - 4 | 219 | 89 | ||
XM_006256065.2 | XP_006256127.1 | 4 - 5 | 334 | 89 | ||
GenBank mRNA | AB000818.1 | - | 2 - 3 | 182 | 89 | |
BC081822.1 | - | 1 - 2 | 752 | 89 | ||
D82069.1 | - | 4 - 5 | 339 | 89 | ||
FM053942.1 | - | 2 - 3 | 171 | 89 | ||
FQ231499.1 | - | 2 - 3 | 182 | 89 | ||
U17919.1 | - | 2 - 3 | 156 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |