Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003974.2 | NP_001003974.1 | 3 - 4 | 652 | 81 | |
GenBank mRNA | BC088220.1 | - | 3 - 4 | 652 | 7384543 | 81 |
FQ229559.1 | - | 3 - 4 | 472 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |