Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024397.2 | NP_077373.2 | 1 - 2 | 182 | 140 | |
XM_006254353.2 | XP_006254415.1 | 1 - 2 | 253 | 140 | ||
XM_006254354.2 | XP_006254416.1 | 1 - 2 | 252 | 140 | ||
XM_006254355.2 | XP_006254417.1 | 1 - 2 | 252 | 140 | ||
XM_006254356.2 | XP_006254418.1 | 1 - 2 | 252 | 140 | ||
XM_006254357.2 | XP_006254419.1 | 1 - 2 | 252 | 140 | ||
XM_006254358.2 | XP_006254420.1 | 1 - 2 | 252 | 140 | ||
XM_006254359.2 | XP_006254421.1 | 1 - 2 | 252 | 140 | ||
XM_006254360.2 | XP_006254422.1 | 1 - 2 | 270 | 140 | ||
GenBank mRNA | AF176784.1 | - | 1 - 2 | 119 | 140 | |
BC129092.1 | - | 1 - 2 | 138 | 7192971 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |