Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236052.2 | XP_006236114.1 | 3 - 4 | 281 | 139 | |
XM_006236053.2 | XP_006236115.1 | 3 - 4 | 263 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |