Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277450.1 | NP_001264379.1 | 8 - 9 | 1205 | 149 | |
NM_019191.2 | NP_062064.1 | 8 - 9 | 1233 | 149 | ||
XM_006254945.2 | XP_006255007.1 | 7 - 8 | 1229 | 149 | ||
XM_006254946.2 | XP_006255008.1 | 8 - 9 | 946 | 149 | ||
GenBank mRNA | AB010147.1 | - | 7 - 8 | 1007 | 149 | |
AB017912.1 | - | 8 - 9 | 1099 | 149 | ||
AF056001.1 | - | 8 - 9 | 1133 | 149 | ||
BC127497.1 | - | 8 - 9 | 1077 | 8364718 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |