Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105939.2 | NP_001099409.1 | 3 - 4 | 260 | 107 | |
NM_001198562.1 | NP_001185491.1 | 3 - 4 | 222 | 107 | ||
NM_001198580.1 | NP_001185509.1 | 3 - 4 | 6000 | 107 | ||
GenBank mRNA | BC166934.1 | - | 3 - 4 | 210 | 6922001 | 107 |
BP494723.1 | - | 3 - 4 | 244 | 107 | ||
FQ221678.1 | - | 3 - 4 | 255 | 107 | ||
FQ222263.1 | - | 3 - 4 | 195 | 107 | ||
FQ231879.1 | - | 3 - 4 | 249 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |