Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031086.2 | NP_112348.2 | 1 - 2 | 155 | 120 | |
GenBank mRNA | CB790907.1 | - | 1 - 2 | 208 | 120 |
Set Membership: |
![]() ![]() ![]() ![]() |