Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031086.2 | NP_112348.2 | 7 - 8 | 814 | 88 | |
XM_006240719.2 | XP_006240781.1 | 5 - 6 | 1100 | 88 | ||
XM_008765045.1 | XP_008763267.1 | 5 - 6 | 546 | 88 | ||
GenBank mRNA | CB580854.1 | - | 2 - 3 | 171 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |