Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107101.1 | NP_001100571.1 | 2 - 3 | 423 | 60 | |
XM_006240716.2 | XP_006240778.1 | 1 - 2 | 204 | 60 | ||
XM_006240717.2 | XP_006240779.1 | 1 - 2 | 202 | 60 | ||
XM_006240718.2 | XP_006240780.1 | 1 - 2 | 202 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |