Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107101.1 | NP_001100571.1 | 6 - 7 | 1092 | 77 | |
XM_006240716.2 | XP_006240778.1 | 5 - 6 | 873 | 77 | ||
XM_006240717.2 | XP_006240779.1 | 5 - 6 | 871 | 77 | ||
XM_006240718.2 | XP_006240780.1 | 5 - 6 | 871 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |