Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106209.1 | NP_001099679.1 | 7 - 8 | 1138 | 114 | |
XM_006227918.2 | XP_006227980.1 | 9 - 10 | 2041 | 114 | ||
XM_008758750.1 | XP_008756972.1 | 6 - 7 | 1290 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |