Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khsrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133602.1 | NP_598286.1 | 11 - 12 | 1082 | 105 | |
XM_006244292.2 | XP_006244354.1 | 11 - 12 | 1220 | 105 | ||
GenBank mRNA | AF308818.1 | - | 11 - 12 | 1082 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |