Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053471.2 | NP_445923.1 | 5 - 6 | 690 | 68 | |
XM_006244348.2 | XP_006244410.1 | 6 - 7 | 1159 | 68 | ||
XM_006244349.2 | XP_006244411.1 | 5 - 6 | 512 | 68 | ||
GenBank mRNA | AB000776.1 | - | 5 - 6 | 690 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |