Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766798.1 | XP_008765020.1 | 2 - 3 | 478 | 78 | |
XM_008766799.1 | XP_008765021.1 | 2 - 3 | 421 | 78 | ||
XM_008766800.1 | XP_008765022.1 | 2 - 3 | 211 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |