Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008882.1 | NP_001008882.1 | 12 - 13 | 1711 | 85 | |
GenBank mRNA | AY321334.1 | - | 12 - 13 | 1711 | 85 | |
BC099224.1 | - | 366 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |