Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3gl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031239.2 | NP_112518.1 | 5 - 6 | 604 | 60 | |
GenBank mRNA | AB008161.1 | - | 5 - 6 | 604 | 60 | |
AF009602.1 | - | 5 - 6 | 589 | 60 | ||
BC070893.1 | - | 5 - 6 | 600 | 7099223 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |