Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mospd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014107.1 | NP_001014129.1 | 3 - 4 | 315 | 118 | |
XM_006257631.2 | XP_006257693.1 | 2 - 3 | 361 | 118 | ||
XM_006257632.2 | XP_006257694.1 | 3 - 4 | 400 | 118 | ||
XM_006257633.2 | XP_006257695.1 | 2 - 3 | 358 | 118 | ||
XM_006257634.2 | XP_006257696.1 | 2 - 3 | 358 | 118 | ||
GenBank mRNA | BC086521.1 | - | 3 - 4 | 315 | 7302680 | 118 |
Gene Symbol: LOC686087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251400.2 | XP_006251462.1 | 1 | 333 | 118 | |
XM_008770405.1 | XP_008768627.1 | 1 | 333 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |