Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053716.1 | NP_446168.1 | 2 - 3 | 396 | 134 | |
GenBank mRNA | AJ005046.1 | - | 2 - 3 | 396 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |