Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hiatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107334.1 | NP_001100804.1 | 4 - 5 | 761 | 132 | |
GenBank mRNA | BC166784.1 | - | 4 - 5 | 754 | 7935371 | 132 |
FQ230882.1 | - | 4 - 5 | 766 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |