Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Habp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079940.1 | NP_001073409.1 | 1 - 2 | 426 | 111 | |
XM_006253494.2 | XP_006253556.1 | 1 - 2 | 481 | 111 | ||
GenBank mRNA | BC129111.1 | - | 1 - 2 | 426 | 7111244 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |