Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138511.1 | NP_612520.1 | 2 - 3 | 557 | 94 | |
GenBank mRNA | L20468.1 | - | 2 - 3 | 557 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |