Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vamp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249016.2 | XP_006249078.1 | 1 - 2 | 142 | 234 | |
GenBank mRNA | FQ228923.1 | - | 1 - 2 | 102 | 234 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |