Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crlf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134465.2 | NP_604460.2 | 4 - 5 | 597 | 103 | |
XM_008769915.1 | XP_008768137.1 | 4 - 5 | 555 | 103 | ||
GenBank mRNA | AF404510.1 | - | 4 - 5 | 540 | 103 | |
BC078740.1 | - | 4 - 5 | 597 | 7108137 | 103 | |
FQ232217.1 | - | 4 - 5 | 583 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |