Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Praf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109013.1 | NP_001102483.1 | 1 - 2 | 331 | 86 | |
GenBank mRNA | FQ213026.1 | - | 1 - 2 | 190 | 86 | |
FQ213646.1 | - | 1 - 2 | 187 | 86 | ||
FQ219908.1 | - | 1 - 2 | 190 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |