Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv39h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106956.1 | NP_001100426.1 | 4 - 5 | 1424 | 71 | |
XM_006256703.2 | XP_006256765.1 | 4 - 5 | 1640 | 71 | ||
XM_006256704.2 | XP_006256766.1 | 4 - 5 | 1216 | 71 | ||
XM_006256705.2 | XP_006256767.1 | 4 - 5 | 1217 | 71 | ||
GenBank mRNA | BC160871.1 | - | 4 - 5 | 991 | 7364965 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |