Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257420.2 | XP_006257482.1 | 1 - 2 | 426 | 67 | |
XM_006257423.2 | XP_006257485.1 | 1 - 2 | 426 | 67 | ||
XM_006257424.2 | XP_006257486.1 | 1 - 2 | 426 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |