Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199115.2 | NP_954546.1 | 4 - 5 | 799 | 84 | |
GenBank mRNA | AY393999.1 | - | 4 - 5 | 659 | 84 | |
BC078944.1 | - | 4 - 5 | 799 | 7109431 | 84 | |
FQ218836.1 | - | 1 - 2 | 69 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |