Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cstf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013161.1 | NP_001013179.1 | 5 - 6 | 1102 | 109 | |
XM_006235681.1 | XP_006235743.1 | 5 - 6 | 1207 | 109 | ||
XM_006235682.1 | XP_006235744.1 | 5 - 6 | 1151 | 109 | ||
XM_006235683.2 | XP_006235745.1 | 5 - 6 | 1234 | 109 | ||
XM_006235684.2 | XP_006235746.1 | 5 - 6 | 1231 | 109 | ||
XM_006235685.2 | XP_006235747.1 | 5 - 6 | 1209 | 109 | ||
XM_006235686.2 | XP_006235748.1 | 5 - 6 | 1206 | 109 | ||
XM_006235687.2 | XP_006235749.1 | 5 - 6 | 1203 | 109 | ||
GenBank mRNA | BC091381.1 | - | 5 - 6 | 1102 | 7320917 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |