Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012623.2 | NP_036755.2 | 3 - 4 | 149 | 97 | |
XM_008762687.1 | XP_008760909.1 | 2 - 3 | 287 | 97 | ||
XM_008762688.1 | XP_008760910.1 | 2 - 3 | 286 | 97 | ||
XM_008762691.1 | XP_008760913.1 | 2 - 3 | 281 | 97 | ||
GenBank mRNA | AY082609.1 | - | 3 - 4 | 149 | 97 | |
BC107560.1 | - | 3 - 4 | 249 | 97 | ||
M81855.1 | - | 3 - 4 | 216 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |