Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191103.1 | NP_001178032.1 | 735 | 89 | ||
XM_008773612.1 | XP_008771834.1 | 840 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |